Methyl-3,4-bis(brommethyl)benzoat 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR3934-1G
OR3934-250MG
APOSOR3934-1GEA
172
EUR
APOSOR3934-1G
APOSOR3934-250MG
Methyl-3,4-bis(brommethyl)benzoat 97%
Methyl-3,4-bis(brommethyl)benzoat
Formel:
C₁₀H₁₀Br₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00219865 CAS-Nummer: 20896-23-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...