2-Methylbenzeneboronic acid-1,3-propanediol ester 95%

Lieferant: Apollo Scientific
Achtung

OR3940-25G OR3940-5G
APOSOR3940-25GEA 352 EUR
APOSOR3940-25G APOSOR3940-5G
2-Methylbenzeneboronic acid-1,3-propanediol ester 95%
2-Methylbenzeneboronic acid-1,3-propanediol ester
Formel: C₁₀H₁₃BO₂
Lagertemperatur: Kühlschrank
MDL: MFCD06659935
CAS-Nummer: 163517-56-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR