3-Boronobenzolsulfonamid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Aminosulphonyl)benzeneboronic acid, 3-Sulphamoylbenzeneboronic acid

OR3945-1G OR3945-250MG
APOSOR3945-1GEA 1320 EUR
APOSOR3945-1G APOSOR3945-250MG
3-Boronobenzolsulfonamid 97%
3-Boronobenzolsulfonamid
Formel: C₆H₈BNO₄S
Molecular Weight: 201,00 g/mol
Schmelzpunkt: 260…266 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06659840
CAS-Nummer: 850568-74-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR