2,5-Dimethylphenylhydraziniumchlorid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(2,5-Dimethylphenyl)hydrazin Hydrochlorid, 2,5-Dimethylphenylhydrazinhydrochlorid, 2,5-Dimethylphenylhydrazin Hydrochlorid
OR3949-100G
OR3949-250G
OR3949-25G
APOSOR3949-100GEA
95.7
EUR
APOSOR3949-100G
APOSOR3949-250G
APOSOR3949-25G
2,5-Dimethylphenylhydraziniumchlorid 97%
2,5-Dimethylphenylhydraziniumchlorid
Formel:
C₈H₁₃ClN₂ Schmelzpunkt: 205 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013382 CAS-Nummer: 56737-78-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...