5-Nitrochinolin
Lieferant: Apollo Scientific
|
Achtung
|
OR3952-100G
OR3952-10G
OR3952-25G
APOSOR3952-100GEA
71.2
EUR
APOSOR3952-100G
APOSOR3952-10G
APOSOR3952-25G
5-Nitrochinolin
5-Nitroquinoline
Formel:
C₉H₆N₂O₂ Molecular Weight: 174,16 g/mol Schmelzpunkt: 71…73 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006790 CAS-Nummer: 607-34-1 EINECS: 210-134-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...