4-(trans-2-Carboxyvinyl)phenylboronsäure 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: (E)-3-(4-Boronophenyl)acrylic acid

OR3953-1G OR3953-5G OR3953-25G
APOSOR3953-1GEA 81.2 EUR
APOSOR3953-1G APOSOR3953-5G APOSOR3953-25G
4-(trans-2-Carboxyvinyl)phenylboronsäure 96%
4-(trans-2-Carboxyvinyl)phenylboronsäure
Formel: C₉H₉BO₄
Molecular Weight: 191,9764 g/mol
Schmelzpunkt: 222…224 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01075704
CAS-Nummer: 159896-15-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR