4-(Methoxycarbamoyl)phenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-(Methoxycarbamoyl)phenyl)boronic acid, 4-(Methoxycarbamoyl)benzeneboronic acid
OR3955-1G
OR3955-5G
APOSOR3955-1GEA
75.4
EUR
APOSOR3955-1G
APOSOR3955-5G
4-(Methoxycarbamoyl)phenylboronsäure 98%
4-(Methoxycarbamoyl)phenylboronsäure
Formel:
C₈H₁₀BNO₄ Schmelzpunkt: 130…136 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115670 CAS-Nummer: 850568-17-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...