3-Chlor-2-methylbenzolsulfonylchlorid
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-Chlor-2-methylbenzolsulfochlorid, 3-Chlor-2-methylbenzolsulfonsäurechlorid, 3-Chlor-2-methylbenzol-1-sulfonylchlorid
OR3972-10G
OR3972-100G
OR3972-25G
APOSOR3972-10GEA
30.6
EUR
APOSOR3972-10G
APOSOR3972-100G
APOSOR3972-25G
3-Chlor-2-methylbenzolsulfonylchlorid
3-Chlor-2-methylbenzolsulfonylchlorid
Formel:
C₇H₆Cl₂O₂S Molecular Weight: 225.09 Schmelzpunkt: 69…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052198 CAS-Nummer: 80563-86-6 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...