3-(Diethylcarbamoyl)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(N,N-Diethylaminocarbonyl)phenylboronic acid, 3-(Diethylcarbamoyl)benzeneboronic acid, (3-(Diethylcarbamoyl)phenyl)boronic acid

OR3978-1G OR3978-5G
APOSOR3978-1GEA 21.9 EUR
APOSOR3978-1G APOSOR3978-5G
3-(Diethylcarbamoyl)phenylboronsäure 98%
3-(Diethylcarbamoyl)phenylboronsäure
Formel: C₁₁H₁₆BNO₃
Molecular Weight: 221.06 g/mol
Schmelzpunkt: 132…136 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04115684
CAS-Nummer: 237413-05-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR