N-(2-Hydroxyethyl)-4-boronobenzolsulfonamid 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-[N-(2-Hydroxyethyl)sulphamoyl]benzeneboronic acid, (4-(N-(2-Hydroxyethyl)sulphamoyl)phenyl)boronic acid
OR3985-100MG
OR3985-250MG
APOSOR3985-100MGEA
21.9
EUR
APOSOR3985-100MG
APOSOR3985-250MG
N-(2-Hydroxyethyl)-4-boronobenzolsulfonamid 95%
N-(2-Hydroxyethyl)-4-boronobenzolsulfonamid
Formel:
C₈H₁₂BNO₅S Molecular Weight: 245.0600 g/mol Schmelzpunkt: 278…284 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD07363750 CAS-Nummer: 850568-77-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...