5-Brom-m-kresol 98%

Lieferant: Apollo Scientific

Synonyme: 3-Bromo-5-methylphenol

OR399005-10G OR399005-5G
APOSOR399005-10GEA 120 EUR
APOSOR399005-10G APOSOR399005-5G
5-Brom-m-kresol 98%
5-Brom-m-kresol
Formel: C₇H₇BrO
Molecular Weight: 187,0300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09038281
CAS-Nummer: 74204-00-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR