4-Brom-2-chloranisol

Lieferant: Apollo Scientific

Synonyme: 4-Bromo-2-chloro-1-methoxybenzene

OR399021-25G
APOSOR399021-25GEA 65.4 EUR
APOSOR399021-25G
4-Brom-2-chloranisol
4-Brom-2-chloranisol
Formel: C₇H₆BrClO
Molecular Weight: 221,4789 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00128076
CAS-Nummer: 50638-47-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR