4-Bromisatosäureanhydrid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Bromo-1H-benzo[d][1,3]oxazine-2,4-dione

OR400015-1G OR400015-25G OR400015-5G
APOSOR400015-1GEA 56.7 EUR
APOSOR400015-1G APOSOR400015-25G APOSOR400015-5G
4-Bromisatosäureanhydrid 97%
4-Bromisatosäureanhydrid
Formel: C₈H₄BrNO₃
Molecular Weight: 242,03 g/mol
Schmelzpunkt: 256 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11113115
CAS-Nummer: 76561-16-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR