4-Methylisatosäureanhydrid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Methyl-1H-benzo[d][1,3]oxazine-2,4-dione

OR400018-1G OR400018-5G
APOSOR400018-1GEA 60.9 EUR
APOSOR400018-1G APOSOR400018-5G
4-Methylisatosäureanhydrid 97%
4-Methylisatosäureanhydrid
Formel: C₉H₇NO₃
Molecular Weight: 177,1568 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03426400
CAS-Nummer: 63480-11-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR