4-Nitroisatosäureanhydrid 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
7-Nitro-1H-benzo[d][1,3]oxazine-2,4-dione
OR400019-1G
OR400019-5G
OR400019-10G
APOSOR400019-1GEA
21.9
EUR
APOSOR400019-1G
APOSOR400019-5G
APOSOR400019-10G
4-Nitroisatosäureanhydrid 95%
4-Nitroisatosäureanhydrid
Formel:
C₈H₄N₂O₅ Molecular Weight: 208,13 g/mol Schmelzpunkt: 253 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00798220 CAS-Nummer: 63480-10-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...