(2-Brom-4-chlorphenyl)essigsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromo-4-chlorophenylacetic acid
OR400058-25G
OR400058-5G
APOSOR400058-25GEA
230
EUR
APOSOR400058-25G
APOSOR400058-5G
(2-Brom-4-chlorphenyl)essigsäure 98%
(2-Brom-4-chlorphenyl)essigsäure
Formel:
C₈H₆BrClO₂ Molecular Weight: 249,4900 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09032951 CAS-Nummer: 52864-56-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...