5,6-Dihydroxyindole 98%

Lieferant: Apollo Scientific
Gefahr

OR40006-1G OR40006-5G
APOSOR40006-1GEA 92.8 EUR
APOSOR40006-1G APOSOR40006-5G
5,6-Dihydroxyindole 98%
5,6-Dihydroxyindole
Formel: C₈H₇NO₂
Molecular Weight: 149,149 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00798933
CAS-Nummer: 3131-52-0
EINECS: 412-130-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR