1-(2-Bromo-6-methoxyphenyl)ethanone

Lieferant: Apollo Scientific

Synonyme: 2-Bromo-6-methoxyacetophenone

OR400097-1G OR400097-5G
APOSOR400097-1GEA 29 EUR
APOSOR400097-1G APOSOR400097-5G
1-(2-Bromo-6-methoxyphenyl)ethanone
1-(2-Bromo-6-methoxyphenyl)ethanone
Formel: C₉H₉BrO₂
Molecular Weight: 229,07 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22573041
CAS-Nummer: 380225-68-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR