Methyl-4-iod-3-nitrobenzoat

Lieferant: Apollo Scientific

Synonyme: 4-Iod-3-nitrobenzoesäuremethylester

OR400138-25G
APOSOR400138-25GEA 127 EUR
APOSOR400138-25G
Methyl-4-iod-3-nitrobenzoat
Methyl-4-iod-3-nitrobenzoat
Formel: C₈H₆INO₄
Molecular Weight: 307,04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08064223
CAS-Nummer: 89976-27-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR