6-Brom-3-coumaranon
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Bromobenzofuran-3(2H)-one
OR40014-5G
OR40014-1G
OR40014-250MG
APOSOR40014-5GEA
317
EUR
APOSOR40014-5G
APOSOR40014-1G
APOSOR40014-250MG
6-Brom-3-coumaranon
6-Brom-3-coumaranon
Formel:
C₈H₅BrO₂ Molecular Weight: 213.0281 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08437617 CAS-Nummer: 201809-69-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...