4-Bromo-5-methyl-2-nitroaniline
Lieferant: Apollo Scientific
OR400179-1G
OR400179-5G
OR400179-250MG
APOSOR400179-1GEA
37.7
EUR
APOSOR400179-1G
APOSOR400179-5G
APOSOR400179-250MG
4-Bromo-5-methyl-2-nitroaniline
4-Bromo-5-methyl-2-nitroaniline
Formel:
C₇H₇BrN₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD16619131 CAS-Nummer: 827-32-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...