(3-Bromphenyl)methylsulfon
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Brom-3-(methylsulfonyl)benzol
OR40020-25G
OR40020-5G
APOSOR40020-25GEA
55.1
EUR
APOSOR40020-25G
APOSOR40020-5G
(3-Bromphenyl)methylsulfon
(3-Bromphenyl)methylsulfon
Formel:
C₇H₇BrO₂S Molecular Weight: 235.1 g/mol Schmelzpunkt: 102…107 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095326 CAS-Nummer: 34896-80-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...