4-Ethinylbenzoesäure 95%

Lieferant: Apollo Scientific
Achtung

OR400230-1G OR400230-25G OR400230-5G
APOSOR400230-1GEA 34.8 EUR
APOSOR400230-1G APOSOR400230-25G APOSOR400230-5G
4-Ethinylbenzoesäure 95%
4-Ethynylbenzoic acid
Formel: C₉H₆O₂
Molecular Weight: 146.1400 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00168819
CAS-Nummer: 10602-00-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR