Methyl-2-methoxy-4-methylbenzoate
Lieferant: Apollo Scientific
Synonyme:
Methyl 2-methoxy-4-methylbenzoate
OR400251-25G
OR400251-5G
APOSOR400251-25GEA
85.7
EUR
APOSOR400251-25G
APOSOR400251-5G
Methyl-2-methoxy-4-methylbenzoate
Methyl-2-methoxy-4-methylbenzoate
Formel:
C₁₀H₁₂O₃ Molecular Weight: 180.2000 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07782177 CAS-Nummer: 81245-24-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...