2,4,6-Trimethoxyphenylboronsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,4,6-Trimethoxybenzeneboronic acid, (2,4,6-Trimethoxyphenyl)metaboric acid, Dihydroxy(2,4,6-trimethoxyphenyl)borane

OR4003-1G OR4003-5G
APOSOR4003-1GEA 89.9 EUR
APOSOR4003-1G APOSOR4003-5G
2,4,6-Trimethoxyphenylboronsäure 97%
2,4,6-Trimethoxyphenylboronsäure
Formel: C₉H₁₃BO₅
Molecular Weight: 212,01 g/mol
Schmelzpunkt: 96…100 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD01114642
CAS-Nummer: 135159-25-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR