Methyl-5-bromo-2-(bromomethyl)benzoate

Lieferant: Apollo Scientific
Achtung

OR400313-1G OR400313-250MG OR400313-5G
APOSOR400313-1GEA 89.9 EUR
APOSOR400313-1G APOSOR400313-250MG APOSOR400313-5G
Methyl-5-bromo-2-(bromomethyl)benzoate
Methyl-5-bromo-2-(bromomethyl)benzoate
Formel: C₉H₈Br₂O₂
Lagertemperatur: Kühlschrank
MDL: MFCD11505949
CAS-Nummer: 79670-17-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR