Methyl-4-brom-2-(brommethyl)benzoat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 4-bromo-2-bromomethylbenzoate
OR400314-1G
OR400314-25G
OR400314-5G
APOSOR400314-1GEA
52.2
EUR
APOSOR400314-1G
APOSOR400314-25G
APOSOR400314-5G
Methyl-4-brom-2-(brommethyl)benzoat
Methyl-4-brom-2-(brommethyl)benzoat
Formel:
C₉H₈Br₂O₂ Schmelzpunkt: 79…80 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD04114319 CAS-Nummer: 78471-43-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...