1-Ethynyl-4-nitrobenzene 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Nitrophenylacetylene
OR400334-1G
OR400334-5G
OR400334-25G
APOSOR400334-1GEA
21.9
EUR
APOSOR400334-1G
APOSOR400334-5G
APOSOR400334-25G
1-Ethynyl-4-nitrobenzene 95%
1-Ethynyl-4-nitrobenzene
Formel:
C₈H₅NO₂ Molecular Weight: 147,13 g/mol Schmelzpunkt: 148…150 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00024794 CAS-Nummer: 937-31-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...