3-Chlor-2-methoxyphenylboronsäure 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-Chloro-2-methoxyphenyl)boronic acid

OR400468-1G OR400468-250MG
APOSOR400468-1GEA 26.1 EUR
APOSOR400468-1G APOSOR400468-250MG
3-Chlor-2-methoxyphenylboronsäure 99%
3-Chlor-2-methoxyphenylboronsäure
Formel: C₇H₈BClO₃
Molecular Weight: 186.4006 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08277278
CAS-Nummer: 179898-50-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR