2-Iodo-4-methoxyaniline 95%

Lieferant: Apollo Scientific

OR400551-1G OR400551-5G
APOSOR400551-1GEA 77 EUR
APOSOR400551-1G APOSOR400551-5G
2-Iodo-4-methoxyaniline 95%
2-Iodo-4-methoxyaniline
Formel: C₇H₈INO
Molecular Weight: 249,05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07776739
CAS-Nummer: 191348-14-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR