1-(5-Bromo-2-nitrophenyl)ethanone

Lieferant: Apollo Scientific

Synonyme: 5’-Bromo-2’-nitroacetophenone

OR400555-5G
APOSOR400555-5GEA 134 EUR
APOSOR400555-5G
1-(5-Bromo-2-nitrophenyl)ethanone
1-(5-Bromo-2-nitrophenyl)ethanone
Formel: C₈H₆BrNO₃
Molecular Weight: 244,0400 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10698995
CAS-Nummer: 41877-24-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR