3-(Dimethylamino)benzaldehyd 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(N,N-Dimethylamino)benzaldehyde

OR400567-250MG OR400567-1G
APOSOR400567-250MGEA 114 EUR
APOSOR400567-250MG APOSOR400567-1G
3-(Dimethylamino)benzaldehyd 95%
3-(Dimethylamino)benzaldehyde
Formel: C₉H₁₁NO
Molecular Weight: 149,1897 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05148649
CAS-Nummer: 619-22-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR