4-Iodo-3-methoxybenzenecarboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Iodo-3-methoxybenzoic acid

OR400602-1G
APOSOR400602-1GEA 48 EUR
APOSOR400602-1G
4-Iodo-3-methoxybenzenecarboxylic acid
4-Iodo-3-methoxybenzenecarboxylic acid
Formel: C₈H₇IO₃
Molecular Weight: 278,05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09753732
CAS-Nummer: 282087-44-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR