4-Brom-2-nitrobenzaldehyd 97%

Lieferant: Apollo Scientific
Achtung

OR400615-25G OR400615-100G
APOSOR400615-25GEA 68.3 EUR
APOSOR400615-25G APOSOR400615-100G
4-Brom-2-nitrobenzaldehyd 97%
4-Bromo-2-nitrobenzaldehyde
Formel: C₇H₄BrNO₃
Molecular Weight: 230,0156 g/mol
Schmelzpunkt: 95…98 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00463687
CAS-Nummer: 5551-12-2
UN: 3077
ADR: 9+EHS,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR