3-Chlor-2-methoxybenzaldehyd

Lieferant: Apollo Scientific

OR400655-1G OR400655-5G
APOSOR400655-1GEA 68.3 EUR
APOSOR400655-1G APOSOR400655-5G
3-Chlor-2-methoxybenzaldehyd
3-Chloro-2-methoxybenzaldehyde
Formel: C₈H₇ClO₂
Molecular Weight: 170,5930 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09999135
CAS-Nummer: 223778-54-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR