4-Brom-1,2-benzoldiol

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromocatechol, 4-Bromo-1,2-dihydroxybenzene

OR400662-1G OR400662-5G OR400662-10G
APOSOR400662-1GEA 26.1 EUR
APOSOR400662-1G APOSOR400662-5G APOSOR400662-10G
4-Brom-1,2-benzoldiol
4-Bromobenzene-1,2-diol
Formel: C₆H₅BrO₂
Molecular Weight: 189,0067 g/mol
Schmelzpunkt: 87 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00869769
CAS-Nummer: 17345-77-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR