3-Methoxy-o-phenylenediamine
Lieferant: Apollo Scientific
|
Achtung
|
OR400685-1G
OR400685-250MG
OR400685-5G
APOSOR400685-1GEA
114
EUR
APOSOR400685-1G
APOSOR400685-250MG
APOSOR400685-5G
3-Methoxy-o-phenylenediamine
3-Methoxy-o-phenylenediamine
Formel:
C₇H₁₀N₂O Schmelzpunkt: 39…40 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08276903 CAS-Nummer: 37466-89-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...