4-Pentylphenylboronsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-n-Pentylbenzeneboronic acid, (4-Pentylphenyl)boronic acid

OR4007-5G OR4007-25G OR4007-100G
APOSOR4007-5GEA 21.9 EUR
APOSOR4007-5G APOSOR4007-25G APOSOR4007-100G
4-Pentylphenylboronsäure 97%
4-Pentylphenylboronsäure
Formel: C₁₁H₁₇BO₂
Molecular Weight: 192,07 g/mol
Schmelzpunkt: 82…84 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00995151
CAS-Nummer: 121219-12-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR