2,6-Dibromopyrazine
Lieferant: Apollo Scientific
OR40072-25G
APOSOR40072-25GEA
105
EUR
APOSOR40072-25G
2,6-Dibromopyrazine
2,6-Dibromopyrazine
Formel:
C₄H₂Br₂N₂ Molecular Weight: 237,882 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD08061601 CAS-Nummer: 23229-25-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...