2-Bromo-5-iodobenzonitrile 98%

Lieferant: Apollo Scientific
Achtung

OR400771-1G OR400771-5G
APOSOR400771-1GEA 30.6 EUR
APOSOR400771-1G APOSOR400771-5G
2-Bromo-5-iodobenzonitrile 98%
2-Bromo-5-iodobenzonitrile
Formel: C₇H₃BrIN
Molecular Weight: 307,91 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09025663
CAS-Nummer: 1252046-13-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR