3-Brom-4-iodanilin 97%
Lieferant: Apollo Scientific
|
Gefahr
|
OR400799-1G
OR400799-5G
APOSOR400799-1GEA
34.8
EUR
APOSOR400799-1G
APOSOR400799-5G
3-Brom-4-iodanilin 97%
3-Brom-4-iodanilin
Formel:
C₆H₅BrIN Molecular Weight: 297,9191 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09753717 CAS-Nummer: 860435-38-7 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...