6-Brom-2,4-dichlorchinazolin
Lieferant: Apollo Scientific
OR400856-5G
APOSOR400856-5GEA
78.3
EUR
APOSOR400856-5G
6-Brom-2,4-dichlorchinazolin
6-Bromo-2,4-dichloroquinazoline
Formel:
C₈H₃BrCl₂N₂ Molecular Weight: 277,93 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09744007 CAS-Nummer: 102393-82-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...