2-Methoxy-3-methylbenzoic acid 95%

Lieferant: Apollo Scientific
Achtung

OR400879-1G OR400879-250MG
APOSOR400879-1GEA 39.3 EUR
APOSOR400879-1G APOSOR400879-250MG
2-Methoxy-3-methylbenzoic acid 95%
2-Methoxy-3-methylbenzoic acid
Formel: C₉H₁₀O₃
Molecular Weight: 166.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00016522
CAS-Nummer: 26507-91-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR