1-(5-Bromo-3-nitropyridin-2-yl)piperazine 95+%

Lieferant: Apollo Scientific

OR40088-1G OR40088-500MG
APOSOR40088-1GEA 538 EUR
APOSOR40088-1G APOSOR40088-500MG
1-(5-Bromo-3-nitropyridin-2-yl)piperazine 95+%
1-(5-Bromo-3-nitropyridin-2-yl)piperazine
Formel: C₉H₁₁BrN₄O₂
Schmelzpunkt: 89…90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD13176633
CAS-Nummer: 1203499-08-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR