6-Brombenzo[b]thiophen

Lieferant: Apollo Scientific

Synonyme: 6-Bromobenzothiophene

OR400926-1G OR400926-5G
APOSOR400926-1GEA 36.4 EUR
APOSOR400926-1G APOSOR400926-5G
6-Brombenzo[b]thiophen
6-Brombenzo[b]thiophen
Formel: C₈H₅BrS
Molecular Weight: 213,0943 g/mol
Schmelzpunkt: 56…60 °C
Lagertemperatur: Kühlschrank
MDL: MFCD11226847
CAS-Nummer: 17347-32-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR