2-Chlor-6-methoxybenzonitril

Lieferant: Apollo Scientific

OR400971-1G OR400971-5G
APOSOR400971-1GEA 92.8 EUR
APOSOR400971-1G APOSOR400971-5G
2-Chlor-6-methoxybenzonitril
2-Chlor-6-methoxybenzonitril
Formel: C₈H₆ClNO
Molecular Weight: 167,5900 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00019743
CAS-Nummer: 6575-10-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR