4-Chlor-2-methoxybenzaldehyd

Lieferant: Apollo Scientific

OR400975-5G
APOSOR400975-5GEA 156 EUR
APOSOR400975-5G
4-Chlor-2-methoxybenzaldehyd
4-Chloro-2-methoxybenzaldehyde
Formel: C₈H₇ClO₂
Molecular Weight: 170,5900 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08703355
CAS-Nummer: 53581-86-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR