α-Brom-2-chlor-4-nitrotoluol
Lieferant: Apollo Scientific
|
Gefahr
|
OR401009-1G
OR401009-250MG
APOSOR401009-1GEA
335
EUR
APOSOR401009-1G
APOSOR401009-250MG
α-Brom-2-chlor-4-nitrotoluol
α-Brom-2-chlor-4-nitrotoluol
Formel:
C₇H₅BrClNO₂ Molecular Weight: 250,48 g/mol Lagertemperatur: Raumtemperatur |
CAS-Nummer:
42533-63-1 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...