2-Ethinylnaphthalin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Ethynyl-naphthalene
OR40103-1G
OR40103-5G
OR40103-250MG
APOSOR40103-1GEA
58
EUR
APOSOR40103-1G
APOSOR40103-5G
APOSOR40103-250MG
2-Ethinylnaphthalin 98%
2-Ethynylnaphthalene
Formel:
C₁₂H₈ Molecular Weight: 152.1919 g/mol Siedepunkt: 110 °C Schmelzpunkt: 42 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00870424 CAS-Nummer: 2949-26-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...