6-Thioguanin
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
6-Mercaptoguanine, 2-Amino-6-purinethiol, 2-Amino-3,7-dihydro-6H-purine-6-thione, 2-Amino-6-mercaptopurine
OR4016-1G
OR4016-5G
OR4016-25G
APOSOR4016-1GEA
21.9
EUR
APOSOR4016-1G
APOSOR4016-5G
APOSOR4016-25G
6-Thioguanin
6-Thioguanin
Formel:
C₅H₅N₅S Molecular Weight: 167,19 g/mol Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00233553 CAS-Nummer: 154-42-7 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...